Learn More
Ace Glass, Inc. O-RING VITON SIZE 011 O-RING VITON SIZE 011
SDP
Catalog No. 50975523
Encompass_Preferred
Size 011 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-523 Supplier Ace Glass, Inc. Supplier No. 785506
Size 011 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.