Learn More
Ace Glass, Inc. O-RING VITON SIZE 018 O-RING VITON SIZE 018
SDP
Catalog No. 50975526
Encompass_Preferred
Size 018 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-526 Supplier Ace Glass, Inc. Supplier No. 785515
Size 018 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.