Learn More
Ace Glass, Inc. O-RING VITON SIZE 021 O-RING VITON SIZE 021
SDP
Catalog No. 50975528
Encompass_Preferred
Size 021 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-528 Supplier Ace Glass, Inc. Supplier No. 785519
Size 021 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.