Learn More
Ace Glass, Inc. O-RING VITON SIZE 114 O-RING VITON SIZE 114
SDP
Catalog No. 50979498
Encompass_Preferred
Size 114 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-979-498 Supplier Ace Glass, Inc. Supplier No. 785522
Size 114 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.