Learn More
Ace Glass, Inc. O-RING VITON SIZE 116 O-RING VITON SIZE 116
SDP
Catalog No. 50975532
Encompass_Preferred
Size 116 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-532 Supplier Ace Glass, Inc. Supplier No. 785526
Size 116 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.