Learn More
Ace Glass, Inc. O-RING VITON SIZE 127 O-RING VITON SIZE 127
SDP
Catalog No. 50975557
Encompass_Preferred
Size 127 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-557 Supplier Ace Glass, Inc. Supplier No. 785576
Size 127 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.