Learn More
Ace Glass, Inc. O-RING VITON SIZE 213 O-RING VITON SIZE 213
SDP
Catalog No. 50979500
Encompass_Preferred
Size 213 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-979-500 Supplier Ace Glass, Inc. Supplier No. 785536
Size 213 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.