Learn More
Ace Glass, Inc. O-RING VITON SIZE 215 O-RING VITON SIZE 215
SDP
Catalog No. 50975537
Encompass_Preferred
Size 215 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-537 Supplier Ace Glass, Inc. Supplier No. 785537
Size 215 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.