Learn More
Ace Glass, Inc. O-RING VITON SIZE 217 O-RING VITON SIZE 217
SDP
Catalog No. 50979501
Encompass_Preferred
Size 217 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-979-501 Supplier Ace Glass, Inc. Supplier No. 785540
Size 217 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.