Learn More
Ace Glass, Inc. O-RING VITON SIZE 219 O-RING VITON SIZE 219
SDP
Catalog No. 50975542
Encompass_Preferred
Size 219 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-542 Supplier Ace Glass, Inc. Supplier No. 785543
Size 219 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.