Learn More
Ace Glass, Inc. O-RING VITON SIZE 221 O-RING VITON SIZE 221
SDP
Catalog No. 50975547
Encompass_Preferred
Size 221 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-547 Supplier Ace Glass, Inc. Supplier No. 785551
Size 221 Viton o-ring. Pack/6. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.