Learn More
Ace Glass, Inc. O-RING VITON SIZE 223 O-RING VITON SIZE 223
SDP
Catalog No. 50975555
Encompass_Preferred
Size 223 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-555 Supplier Ace Glass, Inc. Supplier No. 785574
Size 223 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.