Learn More
Ace Glass, Inc. O-RING VITON SIZE 336 O-RING VITON SIZE 336
SDP
Catalog No. 50975560
Encompass_Preferred
Size 336 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-560 Supplier Ace Glass, Inc. Supplier No. 785582
Size 336 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.