Learn More
Ace Glass, Inc. O-RING VITON SIZE 341 O-RING VITON SIZE 341
SDP
Catalog No. 50975546
Encompass_Preferred
Size 341 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-546 Supplier Ace Glass, Inc. Supplier No. 785550
Size 341 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.