Learn More
Ace Glass, Inc. O-RING VITON SIZE 343 O-RING VITON SIZE 343
SDP
Catalog No. 50975550
Encompass_Preferred
Size 343 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-550 Supplier Ace Glass, Inc. Supplier No. 785566
Size 343 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.