Learn More
Ace Glass, Inc. O-RING VITON SIZE 348 O-RING VITON SIZE 348
SDP
Catalog No. 50975558
Encompass_Preferred
Size 348 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-975-558 Supplier Ace Glass, Inc. Supplier No. 785579
Size 348 Viton o-ring. Pack/3. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.