Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Replacement: O-RINGS, COUPLER, 47MM

Catalog No. 501996880 Shop All Hach Company Products
Encompass

Replacement Coupler O-Rings, 47mm for Xenosep SPE Systems Apparatus.

Catalog No. 50-199-6880 Supplier Hach Company Supplier No. 2943223
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement Coupler O-Rings, 47mm for Xenosep SPE Systems Apparatus.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.