Learn More
World Precision Instrument O-Rings for II-MRBP, Pkg of 50
SDP
Supplier:  World Precision Instrument II5FOR
Encompass
WPI's Mouse and rat tail cuff blood pressure measuring systems for preclincial research, a true turn-key system when it is key critical for determining accurate, consistent blood pressure on mice, rats or any other laboratory animal test subject. A compact, simple yet versatile system that can test from a single up to 200 animals at a time with independent control of each channel. Simple daisy-chaining of systems allow expansion of systems, up to 200 independently, controlled channels. No other system in the world has such vast capabilities as the IITC blood pressure systems. Single animal systems are controlled from the touch screen which allows keying in all necessary test setups. These O-rings are designed for use with the II-MRBP blood pressure system, and they hold the cuff liners on the cuff. Choose either 0.75" or 0.44" O-rings. Both are sold in packages of 50 O-rings.
Catalog No. 50-253-8766