Learn More
Broadley James Corporation Silicone O-Ring, 50/pk, 25 mm DO Sensors , 25 mm pH Housings
Catalog No. NC0684947
Encompass
Broadley-James supplies O-ring replacements for the various sensors and fittings that they manufacture. Use the sizing chart to confirm exact dimensions.25 mm DO Sensors25 mm pH HousingsSize -R15External (replace regularly to ensure the housing or sensor fits securely into the port)
Catalog No. NC0684947 Supplier Broadley James Corporation Supplier No. AGSR1550
Broadley-James supplies O-ring replacements for the various sensors and fittings that they manufacture. Use the sizing chart to confirm exact dimensions.25 mm DO Sensors25 mm pH HousingsSize -R15External (replace regularly to ensure the housing or sensor fits securely into the port)