Learn More
Chemglass Life Sciences #014 Viton O-Rings
SDP
Catalog No. 50873352
Encompass_Preferred
#014 Viton O-Rings. Used with open-top screw caps and threaded outer joints for making grease-free, vacuum tight seals on 14/10 MINUM-WARE glassware components. O-rings are color coded BROWN for easy identification. Supplied in packages of 144 each. See CG-305 series for complete listing of Viton O-Rings.
Catalog No. 50-873-352 Supplier Chemglass Life Sciences Supplier No. MW10008
#014 Viton O-Rings. Used with open-top screw caps and threaded outer joints for making grease-free, vacuum tight seals on 14/10 MINUM-WARE glassware components. O-rings are color coded BROWN for easy identification. Supplied in packages of 144 each. See CG-305 series for complete listing of Viton O-Rings.