Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-(tert-Butyl)-L-serine | 18822-58-7 | 99.2% | 161.20 | 100 G
SDP

Catalog No. 5000451693
Encompass_Preferred

O-(tert-Butyl)-L-serine is a serine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress related tasks and prevent exercise induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-51693 Supplier Medchemexpress LLC Supplier No. HYW010156100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-(tert-Butyl)-L-serine is a serine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress related tasks and prevent exercise induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.