Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-L-serine 97.0+%, TCI America™
SDP

Catalog No. B39171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39171G 1 g
B39175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39171G Supplier TCI America Supplier No. B39171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18822-58-7
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.201
MDL Number MFCD00066088
InChI Key DDCPKNYKNWXULB-YFKPBYRVSA-N
Synonym h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
PubChem CID 7004940
IUPAC Name (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)N

Specifications

Melting Point 226°C
Color White
Quantity 1 g
Formula Weight 161.20
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-L-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.