Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-L-threonine methyl ester hydrochloride, 95%

Catalog No. AAH6352906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6352906 5 g
AAH6352903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6352906 Supplier Thermo Scientific Chemicals Supplier No. H6352906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 71989-43-0 | C9H20ClNO3 | 225.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-43-0
Molecular Formula C9H20ClNO3
Molecular Weight (g/mol) 225.71
MDL Number MFCD00077111
InChI Key SDHKEUUZUMQSAD-HHQFNNIRSA-N
Synonym h-thr tbu-ome.hcl, h-thr tbu-ome hcl, 2s,3r-methyl 2-amino-3-tert-butoxy butanoate hydrochloride, o-tert-butyl-l-threonine methyl ester hydrochloride, o-t-butyl-l-threonine methyl ester hydrochloride, methyl o-1,1-dimethylethyl-l-threoninate hydrochloride, l-threonine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, h-thr but-ome hcl, h-thr t-bu-ome*hcl;, ksc491o2b
PubChem CID 16218568
IUPAC Name methyl (2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)[C@@H](C)OC(C)(C)C

Specifications

Beilstein 4716005
Quantity 5 g
Formula Weight 225.71
Percent Purity 95%
Physical Form Solid
Chemical Name or Material O-tert-Butyl-L-threonine methyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.