Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-L-tyrosine 98.0+%, TCI America™
SDP

Catalog No. B32125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32125G 5 g
B32121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32125G Supplier TCI America Supplier No. B32125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18822-59-8
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD00037202
InChI Key SNZIFNXFAFKRKT-NSHDSACASA-N
Synonym h-tyr tbu-oh, o-tert-butyl-l-tyrosine, o-t-butyl-l-tyrosine, h-l-tyr tbu-oh, tyrosine, o-1,1-dimethylethyl, s-2-amino-3-4-tert-butoxy phenyl propanoic acid, 2s-2-amino-3-4-tert-butoxy phenyl propanoic acid, o-tert.butyl-l-tyrosine, o-t-butyl-tyrosine, tyr but
PubChem CID 7408092
IUPAC Name (2S)-2-azaniumyl-3-[4-(tert-butoxy)phenyl]propanoate
SMILES CC(C)(C)OC1=CC=C(C[C@H]([NH3+])C([O-])=O)C=C1

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 5 g
Formula Weight 237.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.