Learn More
Enzo Life Sciences n-Octyl-beta-D-thioglucopyranoside (5g). CAS: 85618-21-9
SDP
Supplier:  Enzo Life Sciences ALX500009G005
Encompass_Preferred
OTG is a nonionic detergent that may be used to isolate membrane proteins from E. coli without inactivation. Stable in aqueous solution and easily removed by dialysis. Alternative name: n-Octyl-beta-D-thioglucoside, OTG. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline powder. Solubility: Soluble in water (8g/100ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9342