Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine, 95%, pract

Catalog No. AC421120010
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC421120010 1 kg
AC421120250 25 g
AC421121000 100 g
AC421122500 250 g
AC421125000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC421120010 Supplier Thermo Scientific Chemicals Supplier No. FLAC421120010

CAS: 119-93-7 | C14H16N2 | 212.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-93-7
MDL Number MFCD00014773
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
PubChem CID 8413
ChEBI CHEBI:34320
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N

Specifications

Melting Point 125°C to 132°C
Color Beige or Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C14H16N2
Quantity 1 kg
Beilstein 13, IV, 419
Merck Index 15, 9674
Molecular Weight (g/mol) 212.296
Formula Weight 212.29
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material o-Tolidine, (pract), 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-358-0
RTECSNumber DD1225000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.