Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine, MilliporeSigma™ Supelco™

Catalog No. 111020115 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0115 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0115 Supplier MilliporeSigma Supelco Supplier No. 31659100MG
Add to Cart
Add to Cart

o-Tolidine is widely used in the aniline dye industry for the synthesis of different azo dyes.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 119-93-7
Flash Point 100°C - Closed Cup
Physical Form Neat
Linear Formula [-C6H3(CH3)-4-NH2]2
MDL Number MFCD00014773
Melting Point 128°C to 131°C
Synonym 3,3 ′-Dimethylbenzidine; 4,4 ′-Bianisidine
Type Analytical Standard
Quantity 100 mg
Formula Weight 212.29
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.