Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine, Powder, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18604915 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-915 2.5 kg Amber Glass Bottle
18-604-912 25 g Amber Glass Bottle
18-604-913 100 g Amber Glass Bottle
18-604-914 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-915 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T10842.5KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T1084, 119-93-7, C14H16N2

Spectrum™ Chemical o-Tolidine, Powder, Reagent is a commercially important aromatic amine used to make dyes. It is also used in the production of certain elastomers and as a reagent and indicator in analytical forensic and clinical chemistry. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 119-93-7
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
IUPAC Name 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
SMILES CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1

Specifications

CAS Max % 1
Melting Range 125-132°C
Packaging Amber Glass Bottle
Quantity 2.5 kg
Percent Purity 97.5%
Grade Reagent
Physical Form Crystalline Powder or Lumps
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.