Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine, Sigma-Aldrich

Catalog No. 01258809 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-809 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-809 Supplier MilliporeSigma Supelco Supplier No. T853325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

o-Tolidine has also been used to determine the presence of laccase activity in T. versicolor fungal colonies.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-93-7
Molecular Weight (g/mol) 212.29 g/mol

Specifications

Color White
Quantity 25 g
Percent Purity ≥95%
Chemical Name or Material o-Tolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.