Learn More
Medchemexpress LLC o-Tolidine Sulfone c 5g
SDP
Catalog No. 5000205527
Encompass_Preferred
o-Tolidine Sulfone(contains 2 6-Dimethyl isomer) (3 7-Diamino-2 8-dimethyldibenzothiophene sulfone(contains 2 6-Dimethyl isomer)) is a drug intermediate for synthesis of various active compounds
Catalog No. 50-002-05527 Supplier Medchemexpress LLC Supplier No. HYW1103195G
o-Tolidine Sulfone(contains 2 6-Dimethyl isomer) (3 7-Diamino-2 8-dimethyldibenzothiophene sulfone(contains 2 6-Dimethyl isomer)) is a drug intermediate for synthesis of various active compounds