Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC o-Toluic acid | 118-90-1 | 99.8% | 136.15 | 25 G
SDP

Catalog No. 5000402391
Encompass_Preferred

O-toluic acid (2-Methylbenzoic acid) is an organic compound that is benzoic acid with a methyl group substituted at the 2-position. O-toluic acid can act as an exogenous metabolite and holds some chemical research value.

  • Solid appearance.
  • White to off-white color.
  • Chemical formula: C8H8O2.
  • Acts as a xenobiotic metabolite.
  • Derived from an endogenous metabolite.
  • Soluble in DMSO (100 mg/mL, 734.48 mM).
  • Powder stable at -20°C for 3 years, 4°C for 2 years.
  • Solution stable at -80°C for 2 years, -20°C for 1 year.

Catalog No. 50-004-02391 Supplier Medchemexpress LLC Supplier No. 25GHY41494
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-toluic acid (2-Methylbenzoic acid) is an organic compound that is benzoic acid with a methyl group substituted at the 2-position. O-toluic acid can act as an exogenous metabolite and holds some chemical research value.

  • Solid appearance.
  • White to off-white color.
  • Chemical formula: C8H8O2.
  • Acts as a xenobiotic metabolite.
  • Derived from an endogenous metabolite.
  • Soluble in DMSO (100 mg/mL, 734.48 mM).
  • Powder stable at -20°C for 3 years, 4°C for 2 years.
  • Solution stable at -80°C for 2 years, -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.