Learn More
Medchemexpress LLC o-Toluic acid | 118-90-1 | MFCD00002477 | 99.77% | 136.15 | 100 G
SDP
Catalog No. 5000404862
Encompass_Preferred
o-Toluic acid (2-Methylbenzoic acid) is an organic compound that is benzoic acid with a methyl group substituted at the 2-position. It can act as an exogenous metabolite and holds some chemical research value.
  • Organic compound
  • Exogenous metabolite
  • Chemical research value
  • White to off-white solid appearance
  • Soluble in DMSO
Catalog No. 50-004-04862 Supplier Medchemexpress LLC Supplier No. HY41494100G
o-Toluic acid (2-Methylbenzoic acid) is an organic compound that is benzoic acid with a methyl group substituted at the 2-position. It can act as an exogenous metabolite and holds some chemical research value.
  • Organic compound
  • Exogenous metabolite
  • Chemical research value
  • White to off-white solid appearance
  • Soluble in DMSO