Learn More
Medchemexpress LLC O-Toluic acid-13C | 70838-82-3 | 99% | 137.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY41494S1100MG
Encompass_Preferred
o-Toluic acid-13C is the 13C labeled o-Toluic acid, which is a benzoic acid substituted by a methyl group at position 2. It functions as a xenobiotic metabolite and is intended for research use only. This compound is valuable as a tracer and an internal standard in quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-04952