Learn More
Medchemexpress LLC o-Toluic acid-13C | 70838-82-3 | 99% | 137.14 | 5 MG
SDP
Catalog No. 5000404510
Encompass_Preferred
o-Toluic acid-13C is the 13C labeled o-Toluic acid. It is a benzoic acid substituted by a methyl group at position 2, playing a role as a xenobiotic metabolite. This compound is intended for research use only and is not for sale to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during the drug development process.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-04510 Supplier Medchemexpress LLC Supplier No. HY41494S15MG
o-Toluic acid-13C is the 13C labeled o-Toluic acid. It is a benzoic acid substituted by a methyl group at position 2, playing a role as a xenobiotic metabolite. This compound is intended for research use only and is not for sale to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during the drug development process.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.