Learn More
Medchemexpress LLC O-Toluic acid-d7 | 207742-73-2 | 99.5% | 143.19 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY41494S50MG
Encompass_Preferred
O-Toluic acid-d7 is the deuterium labeled o-Toluic acid. It is a benzoic acid substituted by a methyl group at position 2, functioning as a xenobiotic metabolite. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitative analysis during the drug development process. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-04418