Learn More
Medchemexpress LLC O-Toluic acid-d7 | 207742-73-2 | MFCD00216766 | 99.5% | 143.19 | 100 MG
SDP
Catalog No. 5000403691
Encompass_Preferred
O-Toluic acid-d7 is the deuterium labeled form of o-Toluic acid (2-Methylbenzoic acid), which is a benzoic acid with a methyl group at position 2. This compound functions as a xenobiotic metabolite and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-03691 Supplier Medchemexpress LLC Supplier No. HY41494S100MG
O-Toluic acid-d7 is the deuterium labeled form of o-Toluic acid (2-Methylbenzoic acid), which is a benzoic acid with a methyl group at position 2. This compound functions as a xenobiotic metabolite and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.