Learn More
Medchemexpress LLC o-Tolyl-isocyanate-d7 | 1219802-73-9 | C8D7NO | 25 MG
SDP
Catalog No. 5000374339
Encompass_Preferred
o-Tolyl-Isocyanate-d7 is the deuterium labeled o-Tolyl-Isocyanate. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74339 Supplier Medchemexpress LLC Supplier No. 25MGHY151180S
o-Tolyl-Isocyanate-d7 is the deuterium labeled o-Tolyl-Isocyanate. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs