Learn More
Medchemexpress LLC O-tolyl-isocyanated7 | 1219802-73-9 | 99.1% | 140.19 g/mol | C8D7NO | 10 MG
SDP
Catalog No. 5000286751
Encompass_Preferred
o-Tolyl-isocyanate-d7 is the deuterium-labeled analogue of o-tolyl isocyanate developed for use as a tracer or internal standard in quantitative analytical methods. It is formulated for high chemical purity and isotopic enrichment to support accurate, reproducible results in spectroscopy and mass spectrometry workflows.
  • High purity (99.1%) for reliable analytical results.
  • High isotopic enrichment (99.0%) to improve mass-spectrometry tracing accuracy.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS analyses.
  • Colorless to light yellow liquid appearance for straightforward handling.
  • Molecular formula C8D7NO and molecular weight 140.19 g/mol for precise calculations.
Catalog No. 50-002-86751 Supplier Medchemexpress LLC Supplier No. HY151180S10MG
o-Tolyl-isocyanate-d7 is the deuterium-labeled analogue of o-tolyl isocyanate developed for use as a tracer or internal standard in quantitative analytical methods. It is formulated for high chemical purity and isotopic enrichment to support accurate, reproducible results in spectroscopy and mass spectrometry workflows.
  • High purity (99.1%) for reliable analytical results.
  • High isotopic enrichment (99.0%) to improve mass-spectrometry tracing accuracy.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS analyses.
  • Colorless to light yellow liquid appearance for straightforward handling.
  • Molecular formula C8D7NO and molecular weight 140.19 g/mol for precise calculations.