Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-Tolylmagnesium Bromide Solution

Catalog No. 298980100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-898-0100ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 29-898-0100ML Supplier Sigma Aldrich Supplier No. 298980100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

o-Tolylmagnesium bromide solution can be used to prepare:An iridium(I) mesityl complex named Ir(o-tolyl)(CO)(dppe).4-(2-Methylphenyl)-4-oxobutanoic acid, a key intermediate for the synthesis of M1 muscarinic selective agonist AC-42 derivatives.

Specifications

CAS 756525-91-4
Melting Point 217°C to 219°C
Molecular Formula C20H22BrOP
Linear Formula HOCH2CH2P(C6H5)3Br
MDL Number MFCD00011901
Quantity 25 g
Molecular Weight (g/mol) 389.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.