Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-Tolylmagnesium Chloride Solution

Catalog No. 360023800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-002-3800ML 800 mL
36-002-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 36-002-3800ML Supplier Sigma Aldrich Supplier No. 360023800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-Tolylmagnesium Chloride Solution

Specifications

CAS 33872-80-9
Density 0.956 g/mL at 25°C
Molecular Formula C7H7ClMg
Linear Formula CH3C6H4MgCl
MDL Number MFCD00010355
Quantity 800 mL
Synonym 2-Methylphenylmagnesium chloride
Molecular Weight (g/mol) 150.89 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.