Learn More
Medchemexpress LLC O-vanillin (2-hydroxy-3-methoxybenzaldehyde) | 148-53-8 | MFCD00003322 | 25g
SDP
Catalog No. 5000266257
Encompass_Preferred
o-Vanillin (2-Vanillin) is a nature product could be extracted from Vanilla planifolia Pinus koraiensis fruit o-Vanillin is a potent antifungal agent o-Vanillin inhibits the growth of mycelia by disrupting the integrity of cell walls and cell membranes o-Vanillin inhibits Doxorubicin (HY-15142A)- and 4-hydroperoxycyclophosphamide-induced NF- B activation[1][2]
Catalog No. 50-002-66257 Supplier Medchemexpress LLC Supplier No. HYY183225G
o-Vanillin (2-Vanillin) is a nature product could be extracted from Vanilla planifolia Pinus koraiensis fruit o-Vanillin is a potent antifungal agent o-Vanillin inhibits the growth of mycelia by disrupting the integrity of cell walls and cell membranes o-Vanillin inhibits Doxorubicin (HY-15142A)- and 4-hydroperoxycyclophosphamide-induced NF- B activation[1][2]