Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene, ≥99.0% (GC), Solstice

Catalog No. 60048530 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-530 250 mL Glass Bottle
60-048-529 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-530 Supplier Solstice Supplier No. 95662250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 95-47-6 | C8H10 | 106.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -25°C
Appearance Colorless
Density 0.879g/mL at 20°C
Quantity 250 mL
Explosion Limit 0.07
Flash Point 31°C (87.8°F)
Ignition Point 867°F
Packaging Glass Bottle
Refractive Index 1.505
Linear Formula C6H4(CH3)2
Vapor Pressure 0.1 atm (21.1°C), 7mmHg (20°C), 16mmHg (37.7°C)
UN Number UN1307
Beilstein 1815558
Boiling Point 143°C to 145°C
Formula Weight 106.17g/mol
Percent Purity ≥99.0% (GC)
Grade ACS
Chemical Name or Material o-Xylene, 1,2-Dimethylbenzene
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P302 + P352 + P312-P331
DOTInformation class 3 - PG 3 - Xylenes
RTECSNumber 202-422-2
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.