Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene, Reagent Grade, ≥98.0%, Solstice

Catalog No. 60048207 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-207 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-207 Supplier Solstice Supplier No. X10401L

missing translation for 'validMainframeMsg'

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -26°C to -23°C (literature)
CAS Max % 100.0000%
Density 0.879 g/mL at 20 °C(lit.)
Quantity 1 L
Explosion Limit 0.07
Flash Point 30°C
Packaging Glass Bottle
Linear Formula C6H5CF3
Vapor Pressure 16 mmHg (37.7°C), 0.1 atm (21.1°C), 7 mmHg (20°C)
UN Number UN1307
Beilstein 1815558
Boiling Point 143°C to 145°C
Formula Weight 106.17
Percent Purity >98%
Grade Reagent
Chemical Name or Material o-Xylene, Reagent Grade, ≥98.0%
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P302 + P352 + P312-P331
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Xylenes
EINECSNumber 202-422-2
RTECSNumber 202-422-2
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.