Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene 98.0+%, TCI America™
SDP

Catalog No. X0012500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0012500ML 500 mL
X001225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X0012500ML Supplier TCI America Supplier No. X0012500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 95-47-6 | C8H10 | 106.17 g/mol

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -23°C
Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 1307
Boiling Point 145°C
Formula Weight 106.17
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material o-Xylene
EINECSNumber (3)-0003&(3)-0060
RTECSNumber ZE2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.