Learn More
THORLABS INC Ø1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 6", 5 Pack
Catalog No. NC1119238
Encompass
Ø1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 6", 5 Pack
Catalog No. NC1119238 Supplier THORLABS INC Supplier No. TR6P5
Ø1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 6", 5 Pack