Learn More
NEOGEN Modified Tryptone Soy Broth (mTSB), 5 kg
SDP
Catalog No. 502015706
Encompass_Preferred
Modified Tryptone-Soy Broth has emerged as the medium of choice for the enrichment of E. coli O157:H7 in red meats and is incorporated into ISO/TS 13136:2012. This medium is not intended for use in the diagnosis of disease or other conditions in humans. Concern regarding E. coli O157:H7 has grown due to the severity of the disease syndromes caused, and the increase in foodborne infection, so too has the need to optimize methods for its efficient isolation. Symptoms start with severe stomach cramps and watery, bloody diarrhea, and a percentage of individuals infected will develop Haemolytic Uraemic Syndrome (HUS) leading to acute renal failure.For more information visit https://www.neogen.com/solutions/Microbiology/modified-tryptone-soy-broth-mtsb
Catalog No. 50-201-5706 Supplier NEOGEN Supplier No. 700004623
Modified Tryptone-Soy Broth has emerged as the medium of choice for the enrichment of E. coli O157:H7 in red meats and is incorporated into ISO/TS 13136:2012. This medium is not intended for use in the diagnosis of disease or other conditions in humans. Concern regarding E. coli O157:H7 has grown due to the severity of the disease syndromes caused, and the increase in foodborne infection, so too has the need to optimize methods for its efficient isolation. Symptoms start with severe stomach cramps and watery, bloody diarrhea, and a percentage of individuals infected will develop Haemolytic Uraemic Syndrome (HUS) leading to acute renal failure.For more information visit https://www.neogen.com/solutions/Microbiology/modified-tryptone-soy-broth-mtsb