Learn More
Cayman Chemical O1602 Analytical Reference Sta
SDP
Supplier:  Cayman Chemical 100068031 MG
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis; intended for research and forensic applications
Catalog No. 50-288-8772