Learn More
Thermo Scientific O2 CELL SENSOR
Supplier:  Thermo Scientific 290083
Encompass_Preferred
Oxygen Gas Sensors; Thermo Scientific Forma; Temperature Range: -20degree to +45 deg.C, Humidity Range: 0 to 95% RH Non-Condensing, Sensor Life Expectancy: 18 months, Measuring Principle: Galvanic 2-Elec
Catalog No. NC9386487