Learn More
Cayman Chemical O2545 hydrochloride
SDP
Catalog No. 502890379
Encompass_Preferred
A potent water-soluble agonist of CB1 and CB2 receptors with Ki values of 1.5 and 0.32 nM, respectively; when dissolved in saline, is highly efficacious in mouse behavioral models when administered either intravenously or intracerebroventricularly
Catalog No. 50-289-0379 Supplier Cayman Chemical Supplier No. 10009195500 UG
A potent water-soluble agonist of CB1 and CB2 receptors with Ki values of 1.5 and 0.32 nM, respectively; when dissolved in saline, is highly efficacious in mouse behavioral models when administered either intravenously or intracerebroventricularly