Learn More
Cayman Chemical O-2545 (hydrochloride), Purity: ≥98%, CAS: 874745-43-4, 5 mg
SDP
Catalog No. 502891348
Encompass_Preferred
A potent water-soluble agonist of CB1 and CB2 receptors with Ki values of 1.5 and 0.32 nM, respectively; when dissolved in saline, is highly efficacious in mouse behavioral models when administered either intravenously or intracerebroventricularly
Catalog No. 50-289-1348 Supplier Cayman Chemical Supplier No. 100091955 MG
A potent water-soluble agonist of CB1 and CB2 receptors with Ki values of 1.5 and 0.32 nM, respectively; when dissolved in saline, is highly efficacious in mouse behavioral models when administered either intravenously or intracerebroventricularly